Order Entry
Canada
ContactUsLinkComponent
Cadaver Cover for JC101 Body Tray and Frame, Mortech®
Cadaver Cover for JC101 Body Tray and Frame, Mortech®
Catalog # 76498-636
Supplier:  Mopec
undefined
Cadaver Cover for JC101 Body Tray and Frame, Mortech®
Catalog # 76498-636
Supplier:  Mopec
Supplier Number:  JZ012

Specifications

  • Color:
    White
  • Description:
    Cadaver cover for JC101 body tray and frame
  • Cat. no.:
    76498-636

Specifications

About this item

This cadaver cover is designed specifically for use with the JC101 body tray and frame.

  • For use with JC101 Body Tray and frame
  • Color: white

Cadaver Covers allow for the discrete transport of a patient from the hospital room to the mortuary. After placing the body and the frame on the carrier, place cover over the frame. The rectangular shape of the cover conceals the body, giving the illusion of a standard gurney while hiding the cadaver.