12-Place Microscope Slide Holder, Mortech®
Catalog # 76498-606
Supplier: Mopec
Specifications
- Description:12-place microscope slide holder
- Dimensions:3.75×15.25" (95×387 mm)
- Cat. no.:76498-606
Specifications
About this item
Ideal for drying slides (or drying and organizing slides) during frozen sectioning or routine processing.
- Blue polyethylene construction
- Holds 12 slides
Made of durable material, with 12 slide slots designed to hold slides vertically or horizontally.