ContactUsLinkComponent
Bone Dust Collector Hood for Mopec Swordfish, Mortech®
Bone Dust Collector Hood for Mopec Swordfish, Mortech®
Catalog # 76498-410
Supplier:  Mopec
undefined
Bone Dust Collector Hood for Mopec Swordfish, Mortech®
Catalog # 76498-410
Supplier:  Mopec
Supplier Number:  BD147
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Connector hood
  • Cat. no.:
    76498-410
Specifications
About this item
Disposable hoods for the Mopec Swordfish Bone Dust Collector system.
  • Flexible PVC hoods
  • Simple to install over the already mounted blades
Flexible PVC hoods include the hose hook-up for the vacuum system. The bone dust collector hoods will work with all Mopec Swordfish blades, except for the 76 mm large round blade (76498-404).
Packaging: Sold in packs of 10.