Orders LinkContactUsLinkComponent
Instrument Sterilization Trays
Instrument Sterilization Trays
Supplier:   Mopec
Instrument Sterilization Trays
Supplier:  Mopec
Certificates
Instrument Sterilization Trays
Instrument Sterilization Trays
5 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The three-piece tray set can be used for sterilization and decontamination of any pathology dissection instrument. Can also be used in conjunction with Mopec EasySoak Instrument Cleaner – BE046.
  • Stainless steel
  • Resistant to corrosion and oxidation
  • Three pieces include instrument tray, perforated insert tray, and lid
  • Autoclavable
Mopec’s Instrument Sterilization Trays provide secure, efficient organization and sterilization of surgical and laboratory tools. Constructed from durable, corrosion-resistant materials, these trays ensure long-lasting performance and easy cleaning.
Product Family Options
Product Information
  • Capacity
    Material
    Dimensions
    Availability
    Price
  • 76498-236
    Sterilization tray
    Sterilization tray
    22 quarts
    Stainless steel
    20.75×12.75×6”
    Log in for availability
    Log in for availability
    Each
    Each - $900.02
    $900.02
    Each
       $900.02
    Supplier Number: BA054
  • 76549-524
    Sterilization tray
    Sterilization tray
    4 quarts
    Stainless steel
    12.75×10.375×2”
    Log in for availability
    Log in for availability
    Each
    Each - $199.45
    $199.45
    Each
       $199.45
    Supplier Number: BA054-2H
  • 76549-526
    Sterilization tray
    Sterilization tray
    15 quarts
    Stainless steel
    20.75×12.75×4”
    Log in for availability
    Log in for availability
    Each
    Each - $571.12
    $571.12
    Each
       $571.12
    Supplier Number: BA054-4
  • 76549-528
    Sterilization tray
    Sterilization tray
    7 quarts
    Stainless steel
    12.75×10.375×4”
    Log in for availability
    Log in for availability
    Each
    Each - $284.25
    $284.25
    Each
       $284.25
    Supplier Number: BA054-4H
  • 76549-530
    Sterilization tray
    Sterilization tray
    10 quarts
    Stainless steel
    12.75×10.375×6”
    Log in for availability
    Log in for availability
    Each
    Each - $371.60
    $371.60
    Each
       $371.60
    Supplier Number: BA054-6H
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.