ContactUsLinkComponent
Ptec™ Bench Top Ionizer, Removable Emitter Assembly, Transforming Technologies
Ptec™ Bench Top Ionizer, Removable Emitter Assembly, Transforming Technologies
Catalog # 76474-664
Ptec™ Bench Top Ionizer, Removable Emitter Assembly, Transforming Technologies
Catalog # 76474-664
Supplier Number:  IN2000
Specifications
  • Description:
    Bench top ionizer, removable emitter assembly
  • Cat. no.:
    76474-664
Specifications
About this item
The Ptec™ IN2000 AC bench top air ionizer is a small, efficient static elimination device with variable fan speed for protecting electronic components and general static protection.
  • Remove static charges quickly and efficiently
  • Removable emitter point system simplifies service and maintenance
  • Perfect for single station table top, bench, or desk use
  • Balance: ±15 V
  • Small footprint
The IN2000 emitter assembly can be quickly removed and cleaned which eliminates maintenance costs and equipment downtime and ensures the ionizer has optimal static eliminating performance. Using AC corona technology to make the air more conductive, Ptec™ ionizers produce a balanced stream of positive and negative air ions that never need calibration. These devices are used in many industries and manufacturing applications where static electricity is a problem.