Order Entry
Canada
ContactUsLinkComponent
Innovating Science® Soil Macronutrient Test
Innovating Science® Soil Macronutrient Test
Catalog # 470347-446
Supplier:  Ward's Science
CAS Number:  
undefined
Innovating Science® Soil Macronutrient Test
Catalog # 470347-446
Supplier:  Ward's Science
Supplier Number:  IS9761
CAS Number:  

Specifications

  • Chemical method:
    Colorimetry
  • Description:
    Soil macronutrient test kit
  • Environmentally Preferable:
  • Parameters:
    Nitrogen, Potassium, Phosphorus, pH
  • Cat. no.:
    470347-446
  • Application:
    Agriculture
  • No. of Tests:
    50

Specifications

About this item

Easy way to test soil for the macronutrients that grow healthy plants.

  • 50 tests for macronutrient presence in soil samples
  • Aligned with GLOBE protocols

Plants need a variety of nutrients and minerals to grow and maintain overall health. The 3 main nutrients that plants require to survive and thrive are nitrogen, phosphorus, and potassium (also known as NPK). In this activity, students will extract nutrients from a soil sample and perform analyses to determine if nitrogen, phosphorus, and potassium concentration is abundant, adequate, or deficient.

Find problems with plants before they happen.

Delivery: Includes 5 soil collection bottles; 5 plastic scoops; 5 pipettes; 120 ml soil pH indicator; 120 ml nitrogen extraction solution; 30 g nitrogen indicator powder; 120 ml phophorous extraction solution; 30 ml phosphorous reagent; 15 g phosphorouis indicator powder; 120 ml potassium reagent; 15 g potassium indicator powder.