Order Entry
Canada
Orders LinkContactUsLinkComponent
APO-12

APO-12

White crystalline solid
Catalog # CA80057-806
Supplier:   MilliporeSigma
Supplier #:  178377-1GM
APO-12
Catalog # CA80057-806
Supplier:  MilliporeSigma
CAS Number:  
APO-12

White crystalline solid

CAS Number:  
Price
$421.45
Log in for availability
Each (1g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Non-ionic detergent useful in two-phase separation and purification of proteins.

  • Aggregation number 2232, CMC 568 mM.
  • M.W. 246.4.
  • CAS No.: 871-95-4

Documents

Specifications

Specifications

  • Format:
    White crystalline solid

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.