About this item
Anti-Herpes Simplex Virus Type I Antibody was tested in IHC-P with positive control infected tissues. Consult references (Kiernan, 1981: Sheehan & Hrapchak, 1980) for further details on specimen preparation. The user is advised to validate the use of the products with their tissue specimens prepared and handled in accordance with their laboratory practices.
- Reacts with the HSV type 1 specific antigen
- Tested in IHC-P with positive control infected tissues
The herpes simplex (HSV) (also known as cold sore, night fever or fever blister) is a virus that causes a contagious disease. There are two main types of Herpes Simplex Virus (HSV) , 1 and 2; and they belong to a family that includes Epstein-Barr virus (EBV) and Varicella zoster (chicken pox) virus. HSV1 and HSV2 are extremely difficult to distinguish from each other. The HSV-1 strain generally appears in the orafacial organs. HSV2 usually resides in the sacral ganglion at the base of the spine. These viruses have a large DNA genome, capsid, and a lipid bilayer membrane derived from the nuclear membrane of the last host. These viruses are capable of entering a latent phase where the host shows no visible sign of infection. Envelope glycoprotein C (gC) and glycoprotein B (gB) bind to cell surface partials heparin sulfate. Receptor binding protein glycoprotein D (gD) binds to HVEM, nectin-1, and 3-O sulfated heparin sulfate forming complexes with glycoprotein H (gH) and glycoprotein (gL) creating an entry pore for the viral capsid.
Specifications
- Catalog No:
- 76539-422
- CA76539-422
- Antigen symbol:
- HSV1
- HSV1
- Antigen name:
- Herpes Simplex Virus I
- Herpes Simplex Virus I
- Antigen synonyms:
- Simplex|Herpes|HSV|Herpesvirus
- Simplex|Herpes|HSV|Herpesvirus
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Herpes simplex virus
- Herpes simplex virus
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Anti-Herpes Simplex Virus Type I Antibody was produced in rabbit by repeated immunizations with detergent-solubilized herpes simplex virus (HSV) type 1 infected cells.
- Anti-Herpes Simplex Virus Type I Antibody was produced in rabbit by repeated immunizations with detergent-solubilized herpes simplex virus (HSV) type 1 infected cells.
- Format:
- Antiserum
- Antiserum
- Form:
- Liquid
- Liquid
- Purification:
- purified immunoglobulin
- purified immunoglobulin
- Storage buffer:
- 0.01% (w/v) Sodium Azide
- 0.01% (w/v) Sodium Azide
- Storage temperature:
- Store at 2…8 °C
- Store at 2…8 °C
- Shipping temperature:
- Wet Ice
- Wet Ice