Order Entry
Canada
ContactUsLinkComponent
 
Recombinant Oxoguanine Glycosylase
  CA76640-796
 :  
Recombinant Oxoguanine Glycosylase
  CA76640-796
 :  M0464S
 :  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Enzyme type:
    Recombinant
  • Enzyme name:
    OGG
  • Enzyme synonyms:
    Oxoguanine Glycosylase
  • Concentration:
    8,000 units/ml
  • Cat. no.:
    CA76640-796

 

 

Bifunctional DNA glycosylase with DNA N-glycosylase and AP lyase activities

  • Thermostable
  • Isolated from a recombinant source
  • Supplied with 10X Reaction Buffer

Thermostable OGG is an archaeal 8-oxoguanine (8oxoG) DNA glycosylase which acts both as a N-glycosylase and an AP-lyase. The N-glycosylase activity releases the damaged purine (8-oxoguanine) from double stranded DNA, generating an apurinic (AP) site. The AP-lyase activity cleaves 3' to the AP site leaving a 5' phosphate and a 3'-phospho-alpha, beta-unsaturated aldehyde. Unlike some other DNA glycosylases, Thermostable OGG specifically recognizes and cleaves only 8oxoG and no other modified bases.