ContactUsLinkComponent
Innovating Science® STEM Investigations: Make Your Own Liquid Crystal Thermometer
Innovating Science® STEM Investigations: Make Your Own Liquid Crystal Thermometer
  470329-184
undefined
Innovating Science® STEM Investigations: Make Your Own Liquid Crystal Thermometer
  470329-184
 :  IS3454
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    STEM Investigations: Make Your Own Liquid Crystal Thermometer
  • No. of Students:
    8
  • Cat. no.:
    470329-184
Gain an understanding of materials science and liquid crustals and how they can be used in a variety of technologies.
  • Students will form 4 different liquid crystal mixtures that are sensitive to temperature and determine the working temperature range for each of the 4 liquid crystals.
  • Aligned to NGSS Practices
  • Performance expectations: HS-PS2-6, HS-ETS1-2
  • Disciplinary core ideas: ETS1.C
  • Cross cutting concepts include structure and function
  • Engineering practices include obtaining evaluating and communicating information, constructing explanations and designing solutions
Students explore materials science and engineering while learning about an interesting state of matter: liquid crystals. Liquid crystals have an ordered structure like a solid, but are also fluid like a liquid. The molecular-level structure of a liquid crystal can be affected by changes in its environment, such as electric field or temperature, which can lead to changes in its macroscopic properties, such as color. In this experiment, students mix three chemicals in different ratios to form four different liquid crystal mixtures that are sensitive to temperature.
Working in groups, students will determine the working temperature range for each of the four liquid crystal mixtures. As an engineering challenge, the students will then use the information they have gathered to design and construct a thermometer using their liquid crystals. Through this kit, students will gain an understanding of liquid crystals and how they can be used in a variety of everyday technologies. Kit contains enough materials for 8 groups. Teacher's Manual and Student Study Guide copy masters are included.
 : Kit includes instructional manual and SDS booklet.