Order Entry
Canada
Orders LinkContactUsLinkComponent
GelCode™ Blue Stain Reagent, Thermo Scientific

GelCode™ Blue Stain Reagent, Thermo Scientific

GelCode™ Blue Stain Reagent, 500 mL (25 Mini Gels)
Catalog # CAPI24590
Supplier:   Thermo Scientific
Supplier #:  24590
GelCode™ Blue Stain Reagent, Thermo Scientific
Catalog # CAPI24590
Supplier:  Thermo Scientific
CAS Number:  
GelCode™ Blue Stain Reagent, Thermo Scientific

GelCode™ Blue Stain Reagent, 500 mL (25 Mini Gels)

CAS Number:  
Price
$269.22
Log in for availability
Each (500ml)
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

GelCode Blue Stain Reagent is a ready-to-use protein stain based on colloidal coomassie dye G-250 that provides nanogram-level detection in polyacrylamide gels and for in-blot detection.

  • No methanol-acetic acid destaining required
  • Fast, one-step, one-hour staining—ready-to-use stain reagent; band development is visible directly in the staining tray; no background from overnight staining; flexible fixing and washing protocol; no gel shrinkage; stained gel can be dried
  • Water wash increases staining sensitivity—optional step provides crystal-clear gel background and makes weakly stained bands more visible

This unique GelCode Blue Stain Reagent stains only protein and allows bands to be viewed directly in the gel during the one hour gel staining process. Because the destaining step does not fix the protein, GelCode Blue Stain is compatible with mass spectrometry analysis and N-terminal sequence analysis.

 : The 6xHis protein tag staining kit (PI24575) and phosphoprotein staining kit (PI24550) come with the appropriate reagent and control sets, which can be purchased separately.

 : For Research Use Only. Not for use in diagnostic procedures.

Documents

Certificates
Search by Lot Number
Specifications

Specifications

  • Description:
    GelCode™ Blue Stain Reagent
  • Size:
    500 mL (25 Mini Gels)

Specifications