Isolation Gowns, AAMI Level 1, DUKAL™ Corporation
76473-030
: Dukal
- Clothing Size:Universal
- Color:Yellow
- Cat. no.:76473-030
- Packaging:10 per bag, 10 bags per case
Level 1 Isolation Gowns provide a slight barrier against a small amount of fluid penetration.
- Meets AAMI Level 1 requirements for fluid protection
- Slight barrier against a small amount of fluid penetration
- Use in minimal risk situations
Gowns feature elastic wrists with thumbloop to provide a more secure fit under gloves. Tape tab neck with waist ties.