Poly-Coated Isolation Gowns, DUKAL™ Corporation
76473-024
: Dukal
- Color:Yellow
- Cat. no.:76473-024
- Packaging:10 per bag, 5 bags per case
These isolation gowns are poly-coated to provide enhanced fluid protection.
- Made of high quality spunbonded polypropylene
- Poly coated for enhanced fluid protection
Each gown features elastic cuffs with waist and neck tie closures.