You searched for: Tubing
VWR provides a wide selection of tubing varieties that can be suited to any situation, accounting for changing variables such as heat, chemical exposure, and flexibility needs. Tubing can connect between containers in the lab, or it can act as a rigid support structure for a building. The uses are many and varied, and VWR can cover the broad spectrum of needs with its vast collection of tubing, from HPLC and PVC to glass and dialysis tubing.
Masterflex® Low-Density Polyethylene (LDPE) Tubing, Avantor®
Supplier: VWR International
General low pressure and temperature tubing.
Expand 2 Items
Masterflex® I/P® Precision Pump Tubing, Versilon™ A-60-N, Avantor®
Supplier: VWR International
Outlast and outperform other tubing with 10,000 hours of tubing life.
Expand 3 Items
Masterflex® I/P® Precision Pump Tubing, Tygon® S3™ E-LFL, Avantor®
Supplier: VWR International
Long pumping life with superior flexibility reduces downtime and tubing failure.
Expand 3 Items
Masterflex® Transfer Tubing, Flexible Vinyl, Avantor®
Supplier: VWR International
Ideal for general-purpose applications.
Expand 21 Items
Versilon™ 2001 Transfer Tubing
Supplier: VWR International
Ideal for chemical transfer applications.
Expand 8 Items
Hubtron™ Black Pascal Fluoroelastomer Tubing
Supplier: VWR International
Ideal for corrosive chemical transfer.
Expand 3 Items
VWR® Silicone Pump Tubing
Supplier: VWR International
VWR® Silicone pump tubing is extremely flexible and specially formulated for reliable performance in peristaltic pumps.
Expand 7 Items
Masterflex® Transfer Tubing, PharMed® BPT, Avantor®
Supplier: VWR International
Protect your light-sensitive samples.
Expand 16 Items
Masterflex® Transfer Tubing, Versilon™ C-544-A IB, Avantor®
Supplier: VWR International
No plasticizers.
Expand 7 Items
Masterflex® Transfer Tubing, Ester-Based Polyurethane, Avantor®
Supplier: VWR International
Worry-free petroleum-based transfer.
Expand 17 Items
Masterflex® Transfer Tubing, Tygon® Tygon® S3™ E-3603, Avantor®
Supplier: VWR International
Pliable formulation for sharp radius bending.
Expand 30 Items
Masterflex® Transfer Tubing, PVDF, Avantor®
Supplier: VWR International
Fluoropolymer tubing at less cost.
Expand 8 Items
Masterflex® Transfer Tubing, DEHP-Free Flexible PVC, Avantor®
Supplier: VWR International
As flexible as you need it.
Expand 15 Items
Double-Walled Heat-Shrink Tubing, PTFE
Supplier: VWR International
Ideal for chemically inert applications.
Expand 3 Items
Masterflex® Transfer Tubing, Tygon® SE-200, Non-DEHP FEP-Lined, Avantor®
Supplier: VWR International
Keep samples free of odors and taste.
Expand 8 Items
Masterflex® L/S® Precision Pump Tubing, Versilon™ A-60-N, Avantor®
Supplier: VWR International
Well-suited for vacuum/pressure applications.
Expand 6 Items
Masterflex® Ismatec® Microbore Pump Tubing, Platinum-Cured Silicone, Avantor®
Supplier: VWR International
Smooth surface and low protein binding levels.
Expand 5 Items
Masterflex® I/P® Precision Pump Tubing, PharMed® BPT, Avantor®
Supplier: VWR International
Protect your light-sensitive samples.
Expand 3 Items
Masterflex® Ismatec® Microbore Pump Tubing, Tygon® S3™ E-LFL, Avantor®
Supplier: VWR International
Longest life of all Tygon® formulations.
Expand 18 Items
VWR® Red Vacuum Rubber Tubing
Supplier: VWR International
Natural rubber tubing with extra-heavy wall especially recommended for vacuum work.
Expand 9 Items
Masterflex® L/S® Spooled Pump Tubing, Tygon® A-60-F, Avantor®
Supplier: VWR International
Similar to Norprene® but with FDA approval.
Expand 2 Items
Masterflex® L/S® High-Performance Precision Pump Tubing, Versilon™ A-60-N, Avantor®
Supplier: VWR International
Well-suited for vacuum/pressure applications.
Expand 4 Items
Masterflex® L/S® Precision Pump Tubing, PharMed® BPT, Avantor®
Supplier: VWR International
Protect your light-sensitive samples.
Expand 6 Items
Masterflex® I/P® Precision Pump Tubing, Tygon® E-Lab, Avantor®
Supplier: VWR International
No BPA or phthalates—environmentally responsible, more sustainable, and safer.
Expand 3 Items
Masterflex® B/T® PerfectPosition® Pump Tubing, Peroxide-Cured Silicone, Avantor®
Supplier: VWR International
Ensure optimum performance from your Masterflex® pump.
Expand 2 Items
Masterflex® I/P® Precision Pump Tubing, PharmaPure®, Avantor®
Supplier: VWR International
Ultra-low spallation protects fluid purity – ideal for sensitive and costly fluids.
Expand 3 Items
Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, PharMed® BPT, Avantor®
Supplier: VWR International
Designed for optimal operation in your Ismatec pumps.
Expand 18 Items
Masterflex® I/P® High-Performance Precision Pump Tubing, Tygon® A-60-F, Avantor®
Supplier: VWR International
Stay compliant with food grade applications.
Expand 2 Items
Masterflex® L/S® 2-Stop Pump Tubing, Chem-Durance® Bio, Avantor®
Supplier: VWR International
Excellent chemical resistance and long pumping life.
Expand 6 Items
Masterflex® L/S® Precision Pump Tubing, PharmaPure®, Avantor®
Supplier: VWR International
Ultra low spallation protects fluid purity – ideal for sensitive and costly fluids and for filtration applications.