You searched for: Air Samplers
The air can be a serious contamination source, so it is essential for your lab or food processing facility to regularly evaluate the quality of the environment with air samplers. Monitor indoor air quality to ensure viable levels of CO2, humidity, dew point, air temperature, and wet bulb temperature. Sample air by volume for higher accuracy and reproducibility. Graduated glass filter models are designed for use when sampling air volumes at low jet velocity.
Orum International Trio.Bas™ Satellite Kit for ISOLATOR/RABS Units
Supplier: Hardy Diagnostics
A remote aspitating head satellite, available with contact plate or petri plate format, with 5 m long cable, for use with the RABS/ISOLATOR microbial air sampling instrument.
Expand 2 Items
Orum International AIRBIO ONE Kit with Charging Cable, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
The AIRBIO ONE instrument is both a stationary and portable air sampler with one aspirating head.
Expand 4 Items
SP Wilmad-LabGlass Midget Graduated Bubblers, SP Industries
Supplier: SP Industries
Calibrated in 5 mL subdivisions, these graduated, borosilicate glass midget bubblers are used to collect samples of pollutants in air.
Expand 1 Items
Midget Impinger, Ace-Threds, Sintered Glass Filter, Ace Glass Incorporated
Supplier: Ace Glass
This piece is used for sampling air volumes at low jet velocity
Expand 6 Items
Trio.Bas™ Mini Kit, Orum International, Hardy Diagnostics
Supplier: Hardy Diagnostics
Hardy Diagnostics introduces the next generation of microbial air samplers for viable particles.
Expand 1 Items
Impinger, Sherer, Ace Glass Incorporated
Supplier: Ace Glass
This Sherer-type impinger consists of bottle with hex base and 45/50 top joint, and either an SO2 bubbler tube with 25 mm diameter Porosity C (25-50 micron) fritted disc, or an impinger stopper
Expand 5 Items
Impinger, Air Sampling, High Velocity, Ace Glass
Supplier: Ace Glass
This high velocity impinger passes 6L/min
Expand 2 Items
Midget Impinger, Modified, Plain Nozzle, Ace Glass Incorporated
Supplier: Ace Glass
This modified midget impinger has a plain nozzle
Expand 4 Items
SP Wilmad-LabGlass Midget Graduated Impingers, SP Industries
Supplier: SP Industries
Calibrated in 5 mL subdivisions, these graduated, borosilicate glass midget impingers are used to collect samples of pollutants in air.
Expand 1 Items
Trio.Bas™ DUO Kits, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
Hardy Diagnostics introduces the next generation of microbial air samplers for viable particles.
Expand 2 Items
Air Sampling System, C.A.R.B. Octopus Glass, Ace Glass
Supplier: Ace Glass
These vertical systems fit more easily into air sampling stations, trailers and cabinets, and provide a more efficient methodology of sample collection
Expand 6 Items
Midget Impinger, Ace Glass Incorporated
Supplier: Ace Glass
This piece is used for sampling small air volumes at a low jet velocity
Expand 5 Items
Impinger, Air Sampling, Ace Glass Incorporated
Supplier: Ace Glass
This high velocity impinger (Ref AGI-30/AGI-4) passes 12.5L/min (+/-4%, corrected to engineering standard gas conditions) when the pressure drop across the orifice is 41cm Hg. or greater.
Expand 5 Items
Midget Impinger, Modified, Ace Glass Incorporated
Supplier: Ace Glass
This modified midget impinger is used for sampling small air volumes at a low jet velocity
Expand 4 Items
Trio.Bas™ MINI Kit, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
Hardy Diagnostics introduces the next generation of microbial air samplers for viable particles.
Expand 2 Items
Midget Impinger with Ace-Thred Inlet/Outlet, Ace Glass Incorporated
Supplier: Ace Glass
This piece is used for sampling air volumes at low jet velocity
Expand 3 Items
Midget Impinger, Ace Glass
Supplier: Ace Glass
This unique impinger is designed to be more economical, when connecting in series, by eliminating the need for connecting tubes
Expand 1 Items
Graduated Midget Impingers, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
This impinger was designed for the sampling of small air volumes with a low jet velocity
Expand 1 Items
Trio.Bas™ MONO Kits, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
Hardy Diagnostics introduces the next generation of microbial air samplers for viable particles.
Expand 2 Items
MAS-100 NT® & MAS-100 NT® Ex Air Samplers, MilliporeSigma
Supplier: MilliporeSigma
The MAS-100 NT® and MAS-100 NT® Ex portable microbial air samplers are the leading instruments for use in critical environments. These compact yet sophisticated devices are the preferred choice for those demanding the highest quality in microbial air monitoring.
Expand 1 Items
Coriolis® Micro Air Sampler, Bertin Instruments
Supplier: BERTIN CORP
Coriolis® Micro is a portable biological air sampler for indoor or outdoor air bio-contamination monitoring. The equipment concentrates airborne particles in a liquid sample that can be analysed afterwards using microbiological techniques.
Expand 1 Items
Coriolis® COMPACT Air Sampler, Bertin Instruments
Supplier: BERTIN CORP
Portable dry air sampler for bio contamination control.
Expand 1 Items
Expand 2 Items
Aura™ Personal Air Sampler Kits, Restek
Supplier: Restek
New personal air sampling technology designed to meet OSHA and NIOSH requirements for industrial hygiene applications.
Expand 2 Items
Miniature Air Sampling Canisters (Electropolished Stainless Steel and Siltek Treated), Restek
Supplier: Restek
These small canisters are designed for controlled sampling, such as personal air sampling, as an alternative to tube and pump samplers.
Expand 6 Items
Vacuum Bag Sampler, Restek
Supplier: Restek
The model 1062 vacuum bag sampler provides fast sampling with zero cross-contamination.
Expand 1 Items
Coriolis® Air Sampler Accessories, Bertin Instruments
Supplier: BERTIN CORP
Coriolis® air sampler consumables are part of the innovative dry cyclonic technology, which aspirates airborne particles and centrifuges them in a cone.
Expand 7 Items
Passive Air Sampling Kits-Integrated (Stainless Steel and Siltek®-Treated), Restek
Supplier: Restek
Restek’s passive air sampling kit incorporates all the hardware necessary to collect air samples and is easy to assemble for field sampling.
Expand 10 Items
Cleaned Polyurethane Foam (PUF) Cartridges, Restek
Supplier: Restek
These cartridges used for collection of semivolatiles (pesticides, PCBs, PAHs).
Expand 5 Items
Humidification Chamber, Restek
Supplier: Restek
Restek’s humidification chamber is made of acrylic and withstands pressure up to 90 psi.