ContactUsLinkComponent
28 results for Extractors
You searched for: Soxhlet and Soxhlet-Type Extractors
Soxhlet and Soxhlet-Type Extractors
Used when compounds have limited solubility, glass Soxhlet extractors successfully separate lipids from solid material. With the temperature resistance needed to heat solvents, the specialized apparatuses allow vapors to travel up the arm where a thimble blocks solid material from escaping. Designed to have cooling vapor enter the solid contained chamber, percolators circulate the now warm solvent liquid to dissolve desired compounds. Incorporated vapor tubes protect the bulbed siphon tube within Soxhlet extractor systems.
Sort by
Soxhlet Extractor, Glassco
Soxhlet Extractor, Glassco
Supplier: Glassco
Extractor body only.
Expand 2 Items
Loading...
Extractors, Soxhlet with Stopcock, Ace Glass
Extractors, Soxhlet with Stopcock, Ace Glass
Supplier: Ace Glass
This Soxhlet extractor body has a stopcock for withdrawing samples from the extraction chamber
Expand 2 Items
Loading...
WHEATON® Microwave Extraction Tube, DWK Life Sciences
WHEATON® Microwave Extraction Tube, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Manufactured from 33 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Expand 1 Items
Loading...
KIMBLE® Micro Soxhlet Extraction Apparatus, DWK Life Sciences
KIMBLE® Micro Soxhlet Extraction Apparatus, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Soxhlet-type extraction apparatus is for use with extraction thimble, which has a coarse porosity.
Expand 2 Items
Loading...
Extractors, Soxhlet, Ace Glass Incorporated
Extractors, Soxhlet, Ace Glass Incorporated
Supplier: Ace Glass
This extractor body has a ground [ST] 24/40 extractor bottom joint, with ground [ST] outer joints of various sizes at top
Expand 3 Items
Loading...
KONTES® Soxhlet Extraction Apparatus, Kimble Chase, DWK Life Sciences
KONTES® Soxhlet Extraction Apparatus, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Used for the continuous extraction of solids with a suitable solvent
Expand 5 Items
Loading...
Soxhlet and Soxhlet-Type Extractors, Ace Glass
Soxhlet and Soxhlet-Type Extractors, Ace Glass
Supplier: Ace Glass
This Micro Soxhlet Extractor is designed for small chemical applications
Expand 4 Items
Loading...
Extraction Apparatus, Soxhlet with Friedrichs Condenser, Ace Glass
Extraction Apparatus, Soxhlet with Friedrichs Condenser, Ace Glass
Supplier: Ace Glass
The improved design of this Soxhlet extractor with Friedrichs condenser permits greater condensing capacity
Expand 3 Items
Loading...
Soxhlet Extraction Apparatus, DWK Life Sciences
Soxhlet Extraction Apparatus, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Soxhlet extraction apparatus with standard taper ground joints is used for continuous extraction of analytes from a solid into an organic solvent.
Expand 8 Items
Loading...
KIMAX® Extraction Apparatus with Allihn Condenser, Flask, and Soxhlet Extraction Tube, [ST] Joints, Kimble Chase
KIMAX® Extraction Apparatus with Allihn Condenser, Flask, and Soxhlet Extraction Tube, [ST] Joints, Kimble Chase
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Extraction apparatus consists of an Allihn condenser, flask and Soxhlet extraction tube
Expand 9 Items
Loading...
Extraction Apparatus, Soxhlet with Allihn Condenser, Ace Glass
Extraction Apparatus, Soxhlet with Allihn Condenser, Ace Glass
Supplier: Ace Glass
The improved design of this Soxhlet extractor with Allihn condenser permits greater condensing capacity
Expand 4 Items
Loading...
SP Wilmad-LabGlass Soxhlet Extraction Apparatus, Extra-Large, SP Industries
SP Wilmad-LabGlass Soxhlet Extraction Apparatus, Extra-Large, SP Industries
Supplier: SP Industries
This borosilicate glass X-Large Soxhlet extraction apparatus is used for the continuous extraction of solids with a suitable solvent.
Expand 1 Items
Loading...
SP Wilmad-LabGlass Redesigned Extra Large Soxhlet Extraction Apparatus, SP Industries
SP Wilmad-LabGlass Redesigned Extra Large Soxhlet Extraction Apparatus, SP Industries
Supplier: SP Industries
This extra large Soxhlet extraction apparatus has been redesigned to eliminate large diameter Standard Taper joints from the extractor body and condenser.
Expand 1 Items
Loading...
Giant Soxhlet Extraction Apparatus, Ace Glass Incorporated
Giant Soxhlet Extraction Apparatus, Ace Glass Incorporated
Supplier: Ace Glass
This apparatus comes complete with a bulb-type condenser and one flask
Expand 10 Items
Loading...
SP Wilmad-LabGlass Soxhlet Extraction Apparatus, SP Industries
SP Wilmad-LabGlass Soxhlet Extraction Apparatus, SP Industries
Supplier: SP Industries
These borosilicate glass Soxhlet extraction apparatus are used for the continuous extraction of solids with a suitable solvent
Expand 4 Items
Loading...
KIMBLE® Soxhlet Extraction Apparatus with Enlarged Vapor Tube, DWK Life Sciences
KIMBLE® Soxhlet Extraction Apparatus with Enlarged Vapor Tube, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Soxhlet-type extraction apparatus with a bulb-type Allihn condenser, used for the continuous extraction of solids with a suitable solvent.
Expand 2 Items
Loading...
KIMBLE® Soxhlet Extraction Body, DWK Life Sciences
KIMBLE® Soxhlet Extraction Body, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Extractor with PTFE stopcock outlet for periodic solvent removal.
Expand 1 Items
Loading...
SP Wilmad-LabGlass Fat Extraction Flasks, SP Industries
SP Wilmad-LabGlass Fat Extraction Flasks, SP Industries
Supplier: SP Industries
These borosilicate glass extraction apparatus are used to measure the fat content in dairy and other food products.
Expand 1 Items
Loading...
Kimble Chase Mojonnier Fat Extractor Flask
Kimble Chase Mojonnier Fat Extractor Flask
Supplier: DWK Life Sciences (KIMBLE)
Primarily used to determine fat content in dairy products but may also be used for other food products.
Expand 1 Items
Loading...
Synthware Soxhlet Extractors, Kemtech America
Synthware Soxhlet Extractors, Kemtech America
Supplier: Kemtech America
Extractor body only.
Expand 3 Items
Loading...
Soxhlet Extractor, Chemglass
Soxhlet Extractor, Chemglass
Supplier: Chemglass
Micro size soxhlet extractor for use with 10×50 mm paper or glass thimbles.
Expand 1 Items
Loading...
Eisco Soxhlet Extraction Apparatus
Eisco Soxhlet Extraction Apparatus
Supplier: Wards
Soxhlet extraction apparatus consisting of boiling flask, extractor, and condenser, all made from high quality borosilicate glass.
Expand 4 Items
Loading...
Borosil® Complete Soxhlet Extraction Apparatus with Allihn Condenser and Flask, Foxx Life Sciences
Borosil® Complete Soxhlet Extraction Apparatus with Allihn Condenser and Flask, Foxx Life Sciences
Supplier: Foxx Life Sciences
Borosil® Soxhlet extraction apparatus with Allihn condenser is used for the separation of an extraction from a liquid or solid state and is ideal for laboratory scale refluxing.
Expand 6 Items
Loading...
PYREX® Soxhlet Extraction Tubes, [ST] Joints, Corning
PYREX® Soxhlet Extraction Tubes, [ST] Joints, Corning
Supplier: Corning
PYREX®, borosilicate glass.
Expand 4 Items
Loading...
Soxhlet Extraction Apparatus, Chemglass
Soxhlet Extraction Apparatus, Chemglass
Supplier: Chemglass
Used for continuous extraction of solids with a suitable solvent
Expand 6 Items
Loading...
PYREX® Extraction Apparatus, with Friedrichs Condenser, [ST] Joints, Corning
PYREX® Extraction Apparatus, with Friedrichs Condenser, [ST] Joints, Corning
Supplier: Corning
An efficient extraction apparatus for all general purposes
Expand 2 Items
Loading...
PYREX® Extraction Apparatus with Allihn Condenser, Flask, and Soxhlet Extraction Tube, [ST] Joints, Corning
Expand 4 Items
Loading...
Soxhlet Extractors, ChemScience, SATI International Scientific
Soxhlet Extractors, ChemScience, SATI International Scientific
Supplier: CHEM SCIENCE INC CA
Soxhlet Extractors, ChemScience, SATI International Scientific
Expand 3 Items
Loading...
Sort by
Recommended for You