Order Entry
Canada
ContactUsLinkComponent
Eisco® Accessories Set for Linear Air Track
Eisco® Accessories Set for Linear Air Track
Catalog # 470224-380
Supplier:  Wards
undefined
Eisco® Accessories Set for Linear Air Track
Catalog # 470224-380
Supplier:  Wards
Supplier Number:  PH0362BACC

Specifications

  • Description:
    Accessories set for linear air track
  • Cat. no.:
    470224-380
  • Supplier No.:
    PH0362BACC

Specifications

About this item

Accessories for demonstrating properties and laws of motion using linear air tracks.

  • Complete accessory pack for Eisco labs linear air track systems
  • Contains all components required for using the linear air track
  • Use for studying conservation of energy or momentum, elastic and inelastic collisions, Newton's second law, a many other topics
  • Air track not included

The linear air track (not included) provides a nearly frictionless surface with which one can study a multitude of physical processes in ideal conditions.

The air track gliders float on a cushion of air pushed out through small holes in the top of the air track. Using the many included accessories in this kit, experiments can be performed exploring conservation of energy, conservation of momentum, elastic and inelastic collisions, Newton's second law, among many other topics.

Delivery: This accessory pack contains all of the components necessary for use with Eisco labs linear air track apparatus (not included; 470157-654, 470157-664). It includes 2 sliders, 10 bolt and wing nuts, 4 spring attachments, 4 string attachments, 4 springs, 2 velcro attachment pairs, 2 pulleys with strings, 4 slotted masses with 1 hanger, 8 photogate flags, 8 risers, 6 additional masses, and 2 photogate arms.