About this item
This premium HyCloneTM Fetal Bovine Australian-origin Serum is well-characterized, triple 100 nm filtered, virus-tested to 9 CFR standard, and traceability-certified (ISIA, Oritain).
- Benefit from Australian husbandry methods and certified traceability
- Assayed for a variety of critical components and biochemical characteristics
- Certificate of analysis confirms ≤25 EU/ml endotoxin
- Serial 100 nm pore size-rated filtering for reliable microbial depletion
- Gamma rradiated and heat-inactivated FBS options also available
- Broad virus panel testing in accordance with 9 CFR 113.53
HyClone Characterised FBS is an excellent alternative to more expensive defined sera, and meets the requirements of most discriminating FBS users. The final serum is assayed for important proteins, hormones, growth factors, metabolites, and nutrients that support healthy cell cultures. Assay results are included in the certificate of analysis and biochemical assay list. Additional testing, including EMEA-conforming testing, is also available.
HyClone Characterised FBS is triple-filtered through serial 100 nm (0,10 μm) pore size-rated filters, which has become an industry standard for cell culture grade FBS. Before dispensing, each lot of serum is pooled using true pool technology to minimise variability from bottle to bottle.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA76324-882
- CA76324-928
- Origin:
- Animal
- Animal
- Endotoxin:
- ≤25 EU/ml
- ≤25 EU/ml
- Hemoglobin:
- ≤25 mg/dl
- ≤25 mg/dl
- Storage temperature:
- <–10 °C
- <–10 °C