Specifications
- Description:Linearity FLQ TDM for Beckman AU
- Cat. no.:76492-634
- Volume:4 ml
- Includes:5 vials
Specifications
About this item
The Audit® MicroControlsTM Linearity FLQ TDM for Beckman AU is intended to simulate human patient samples for use in determining linearity, calibration verification, and the verification of reportable range for the following analytes: Acetaminophen, Amikacin, Carbamazepine, Digoxin, Gentamicin, Lidocaine, NAPA, Phenobarbital, Phenytoin, Primidone, Procainamide, Quinidine, Salicylate, Theophylline, Tobramycin, Valproic Acid, Vancomycin.