Specifications
- Description:Linearity FLQ urine/fluids chemistry for Beckman AU
- Cat. no.:76492-630
- Volume:3 ml
- Includes:10 vials
Specifications
About this item
The Audit® MicroControlsTM Linearity FLQ Urine/Fluids Chemistry for Beckman AU is intended to simulate human patient samples for use in determining linearity, calibration verification, and the verification of reportable range for the following analytes: Amylase, Calcium, Chloride, Creatinine, Glucose, Magnesium, Microalbumin, Microprotein, Potassium, Sodium, Phosphorous, Urea Nitrogen, Uric Acid.