TempliPhi™ Kits efficiently prepare micrograms of circular DNA from picogram input material. The DNA templates are prepared by rolling circle amplification (RCA) using bacteriophage Phi29 DNA polymerase. TempliPhi™ uses an isothermal method for the exponential amplification of circular DNA. Phi29 DNA polymerase is active at 30 °C, enabling amplification to be performed at this temperature without the need for thermal cycling. The TempliPhi™ protocol requires less than 20 minutes of hands-on time to amplify 96 samples from bacterial colonies.
- Prepare circular DNA templates for cycle sequencing, cloning, and transformation in 4 to 6 hours
- Generate microgram quantities of template DNA from picogram amounts of starting material
- Use amplified DNA directly for cycle sequencing without purification
- Amplify DNA from bacterial or M13 liquid cultures, colonies, plaques, glycerol stocks, or purified circular (plasmid or M13) DNA
- Simple protocol reduces time, labour, and consumables needed for template preparation, and workflow enables easy automation
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Storage conditions:Denature buffer and control DNA at -70°C or -15 to -30°C. TempliPhi premix at -70°C (do not store TempliPhi premix at -15 to -30°C). Thaw premix on ice, maintain at 0-4°C during handling (do not allow premix to warm above 4°C prior to amplification)