ContactUsLinkComponent
UV-C Indicator Card (Double-Sided)
UV-C Indicator Card (Double-Sided)
Catalog # 470334-106
Supplier:  Ward's
undefined
UV-C Indicator Card (Double-Sided)
Catalog # 470334-106
Supplier:  Ward's
Supplier Number:  B1450-IC
Specifications
  • Description:
    UV-C Indicator Card (Double-Sided)
  • Size:
    Pack of 25
  • Cat. no.:
    470334-106
Specifications
About this item
Indicator cards and stickers allow for quick determination of UV-C exposure.
  • Color change clearly indicates UV-C exposure
  • Pack of 25
  • Double-sided