Order Entry
Canada
ContactUsLinkComponent
Corning® Ultra-Low Attachment 25 cm² Rectangular Canted Neck Cell Culture Flask with Vent Cap, Corning
Corning® Ultra-Low Attachment 25 cm² Rectangular Canted Neck Cell Culture Flask with Vent Cap, Corning
Catalog # 76441-540
Supplier:  Corning
   Low Manufacturing Impact
CAS Number:  
undefined
Corning® Ultra-Low Attachment 25 cm² Rectangular Canted Neck Cell Culture Flask with Vent Cap, Corning
Catalog # 76441-540
Supplier:  Corning
Supplier Number:  4616
   Low Manufacturing Impact
CAS Number:  

Specifications

  • Description:
    Corning® ultra-low attachment 25 cm² rectangular canted neck cell culture flask with vent cap; Cell Growth Area: 25 cm² 
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    RECs, ISO 14001 - Oneonta
  • Cat. no.:
    76441-540
  • Volume:
    5 - 7.5 ml

Specifications

About this item

Features a covalently bound hydrogel layer that effectively inhibits cellular attachment and minimizes protein absorption, enzyme activation, and cellular activation.

  • Surface is non-cytotoxic, biologically inert, and nondegradable
  • Lot numbers for ease in traceability
  • 100% integrity tested
  • Sterilized by gamma irradiation
  • Nonpyrogenic

The Corning vessels feature a biologically inert ultra-low attachment surface that minimizes cell attachment and promotes the formation of 3D multicellular spheroids employed in cancer research, stem cell biology, and drug screening.

Minimizes cell attachment, protein absorption, and enzyme activation.