VacElut Cartridge Manifolds, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
VacElut cartridge manifolds enable simultaneous use of multiple Bond Elut cartridges.
- Precise vacuum settings
- Increased productivity/sample throughput
- Eliminate cross contamination with disposable needles
- Rugged, reliable construction
- Transparent glass base allows observation of the whole collection operation
- Simple vacuum adjustment
The corrosion-resistant VacElut cartridge manifolds allow simultaneous processing of multiple Bond Elut cartridges. The clear glass base allows observation of the entire sample collection process. Its compact, linear design requires little bench space.
These manifolds are also able to accommodate larger 16×150 mm test tubes. These collection vessels can be used in combinatorial chemistry applications using large boiling tubes for collection of purified synthesis mixtures. They can be used for any SPE extraction where an elution volume greater than 10 ml is required.
Customers Who Bought This Also Bought