NEBNext® Ultra II Directional RNA Library Prep with Sample Purification Beads, New England Biolabs
Supplier: New England Biolabs (NEB)
Certificates
About this item
The Ultra II Directional RNA Library Prep Kit for Illumina delivers significantly increased sensitivity and specificity from your RNA-seq experiments, from ever-decreasing amounts of input RNA. In conjunction with ribosomal RNA (rRNA) depletion or poly(A) enrichment, the kit enables the production of high quality libraries from 5 ng or 10 ng of Total RNA, respectively, up to 1 µg.
- Generate the highest yields of high quality libraries, with a broad range of input amounts
- Increase the complexity and transcript coverage of your libraries
- Optimize your time with streamlined workflows, reduced hands-on time, and automation compatibility
- Rely on robust performance, even with low quality RNA, including FFPE
- Enjoy the flexibility and reliability of the gold standard SPRIselect beads
The NEBNext Ultra II Directional RNA Library Prep Kit derives its directionality from the 'dUTP' method for strand-specificity, with proven superiority for this application. For convenience, this kit includes SPRISelect beads.
Specifications
- Application:
- Environmentally Preferable:
Specifications