NEBNext® Ultra II Directional RNA Library Prep Kit for Illumina, New England Biolabs
103501-462
:
- Description:NEBNext® Ultra II directional RNA library prep kit for illumina
- Cat. no.:103501-462
- No. of Reactions:96
The Ultra II Directional RNA Library Prep Kit for Illumina delivers significantly increased sensitivity and specificity from your RNA-seq experiments, from ever-decreasing amounts of input RNA. In conjunction with ribosomal RNA (rRNA) depletion or poly(A) enrichment, the kit enables the production of high quality libraries from 5 ng or 10 ng of Total RNA, respectively, up to 1 µg. The NEBNext Ultra II Directional RNA Library Prep Kit derives its directionality from the 'dUTP' method for strand-specificity, with proven superiority for this application.
- Generate the highest yields of high quality libraries, with a broad range of input amounts
- Increase the complexity and transcript coverage of your libraries
- Optimize your time with streamlined workflows, reduced hands-on time, and automation compatibility
- Rely on robust performance, even with low quality RNA, including FFPE