Order Entry
Canada
ContactUsLinkComponent
NEBNext® Multiplex Small RNA Library Prep Kit for Illumina (Index Primers 1 to 48), New England Biolabs
NEBNext® Multiplex Small RNA Library Prep Kit for Illumina (Index Primers 1 to 48), New England Biolabs
Catalog # CA103258-562
CAS Number:  
undefined
NEBNext® Multiplex Small RNA Library Prep Kit for Illumina (Index Primers 1 to 48), New England Biolabs
Catalog # CA103258-562
Supplier Number:  E7560S
CAS Number:  

Specifications

  • Application:
  • Description:
    NEBNext® Multiplex small RNA library prep kit for illumina (index primers 1-48)
  • Environmentally Preferable:
  • Cat. no.:
    CA103258-562
  • No. of Reactions:
    96

Specifications

About this item

The NEBNext Multiplex Small RNA Library Prep Kit for Illumina (Index Primers 1 to 48) contains the adaptors, primers, enzymes and buffers required to convert small RNAs into indexed libraries for next-generation sequencing on the Illumina platform. The novel workflow has been optimized to minimize adaptor-dimers, while producing high-yield, high-diversity libraries.

  • Minimized adaptor-dimer contamination
  • High yields
  • Input RNA can be Total RNA
  • Suitable for methylated small RNAs (e.g., piRNAs) as well as unmethylated small RNAs