Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Concentration:10X
- Description:rCutSmart™ Buffer
- Size:5 ml
- Cat. no.:CA76486-068
Specifications
About this item
New England Biolabs provides a color-coded 10X NEBuffer with each restriction endonuclease to ensure optimal (100%) activity. Most of our enzymes are supplied with one of four standard NEBuffers.
- Contains recombinant Albumin
- Set contains 1.25 ml vial of each buffer
These buffers contain Recombinant Albumin (NEB #B9200). Over 215 restriction enzymes are 100% active in rCutSmart Buffer, making it significantly easier to set up your double digest reactions. Since rCutSmart Buffer includes Recombinant Albumin, there are also fewer tubes and pipetting steps to worry about. Additionally, many DNA modifying enzymes are 100% active in rCutSmart Buffer, eliminating the need for subsequent purification.