HardyDisk™, 15 µg (CLR-15) AST Clarithromycin
Supplier: HARDY DIAGNOSTICS CA
Certificates
About this item
HardyDisk™ AST's are impregnated paper disks used for Antimicrobial Susceptibility Testing (AST); also known as disk diffusion or Kirby-Bauer testing.
Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing.
Delivery information: Freeze on arrival.
Frequently Bought Together