DuoPress™ Wringer for Flat Mops, Vileda Professional®
76486-080
- Color:Blue
- Description:Wringer for flat mops
- Environmentally Preferable:
- Cat. no.:76486-080
Trolleys and accessories that meet the highest cleanroom standards.
- For use on the CE Bucket Trolley with the bucket method
- 100% polypropylene chassis and stainless steel lever
- Effectively wrings CE Duo® mops
- Ergonomic lever for maximum pressure with minimal effort
- Base of press support dropped 10 mm to create additional space for mops; making it easier to wring
The CE Trolley range has been specifically designed for use in controlled environments where maximum contamination control is needed. This DuoPress™ wringer is part of the CE Bucket Trolley line for wringing of flat mops.