To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:12 per pack
- Description:Prepared media (SabDex (Sabouraud Dextrose) agar, USP)
- Size:250 ml
- Minimum Order Quantity:4 packs of 12
- Cat. no.:76339-812
- Fill Volume:100 ml
- Type:Square polycarbonate bottle
Specifications
About this item
Sabouraud dextrose agar is recommended for the isolation, cultivation, and maintenance of non-pathogenic and pathogenic species of fungi and yeasts, and meet the harmonized USP standards for the microbial examination of non sterile products.