Tryptic Soy Agar (TSA) with Lecithin and Tween® 80, Hardy Diagnostics
Catalog #: 76339-810
Supplier: Hardy Diagnostics
CAS Number:
Specifications
- Packaging:12 per pack
- Description:Prepared media (Tryptic soy agar (TSA) with Lecithin and Tween® 80)
- Size:240 ml
- Cat. no.:76339-810
- Fill Volume:100 ml
- Type:Boston round bottle
Specifications
About this item
For use as a general growth medium for the isolation and cultivation of microorganisms. This medium is also recommended for use in the cultivation, storage, maintenance, and transportation of pure cultures of microorganisms.
- A crystal clear glass construction provides complete visibility
- Features a streamlined design with no handle, making it easier to store and clean
- Easy-to-remove, perforated, shrink wrap ensures product integrity
Customers Who Bought This Also Bought