Order Entry
Canada
ContactUsLinkComponent
CompactDry™ Salmonella (SL), Hardy Diagnostics
CompactDry™ Salmonella (SL), Hardy Diagnostics
Catalog # CA10789-458
CAS Number:  
undefined
CompactDry™ Salmonella (SL), Hardy Diagnostics
Catalog # CA10789-458
Supplier Number:  54085
CAS Number:  

Specifications

  • Description:
    CompactDry™ Salmonella (SL), mini plates for colony counts
  • Size:
    240 plates/pack
  • Storage temperature:
    2 to 30 °C away from direct light
  • Cat. no.:
    CA10789-458
  • Supplier No.:
    SL100

Specifications

About this item

CompactDry™ SL is a ready-to-use test method recommended for the determination of total aerobic bacterial counts.

  • Sturdy frame, stackable packaging
  • Self-diffusing media
  • Ability to subculture without damage to colonies
  • No leakage
  • Easy to label

Each cassette comes with 4 connected plates that can be easily snapped apart to support a variety of dilution scenarios.

Caution: Stored at room temperature vs the competitor products with storage at temperatures less than or equal to 8 °C.