ContactUsLinkComponent
CompactDry™ Yeast/Mold Rapid (YMR), Hardy Diagnostics
CompactDry™ Yeast/Mold Rapid (YMR), Hardy Diagnostics
Catalog # CA76415-000
CAS Number:  
undefined
CompactDry™ Yeast/Mold Rapid (YMR), Hardy Diagnostics
Catalog # CA76415-000
Supplier Number:  54084
CAS Number:  
Specifications
  • Description:
    CompactDry™ Yeast/Mold Rapid (YMR) cassettes for colony counts 
  • Size:
    240 plates/pack
  • Cat. no.:
    CA76415-000
Specifications
About this item
CompactDry™ YMR is a ready-to-use test method recommended for the determination of total aerobic bacterial counts.
  • Sturdy frame, stackable packaging
  • Self-diffusing media
  • Ability to subculture without damage to colonies
  • No leakage
  • Easy to label
Each cassette comes with 4 connected plates that can be easily snapped apart to support a variety of dilution scenarios.
Caution: Stored at room temperature vs the competitor products with storage at temperatures less than or equal to 8 °C.