CompactDry™ Yeast/Mold (YM), Hardy Diagnostics
Catalog # CA10145-972
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Specifications
- Description:CompactDry™ Yeast/Mold (YM) cassettes
- Size:240 plates/pack
- Storage temperature:2 to 30 °C away from direct light
- Cat. no.:CA10145-972
- Supplier No.:YM100
Specifications
About this item
CompactDry™ YM is a ready-to-use test method recommended for the determination of total aerobic bacterial counts.
- Sturdy frame, stackable packaging
- Self-diffusing media
- Ability to subculture without damage to colonies
- No leakage
- Easy to label
CompactDry™ is a ready-to-use system that reduces the time needed to perform microbial testing on food, cannabis, meat, cosmetics, raw materials and more.
Each cassette comes with 4 connected plates that can be easily snapped apart to support a variety of dilution scenarios.
Caution: Stored at room temperature vs the competitor products with storage at temperatures less than or equal to 8 °C.