ContactUsLinkComponent
Cuvette Swabs, 6" Spatula Foam Swab with Black Polypropylene Handle, Hardy Diagnostics
Cuvette Swabs, 6" Spatula Foam Swab with Black Polypropylene Handle, Hardy Diagnostics
Catalog # 76320-912
Supplier:  Hardy Diagnostics
undefined
Cuvette Swabs, 6" Spatula Foam Swab with Black Polypropylene Handle, Hardy Diagnostics
Catalog # 76320-912
Supplier:  Hardy Diagnostics
Supplier Number:  QS1001
Specifications
  • Description:
    Spatula foam swab
  • Cat. no.:
    76320-912
  • Packaging:
    100 per pack
Specifications
About this item
Specifically designed for QuickSlide™ automated stainers, the foam-tipped swab is ideal for maintaining a clean cuvette, free of stain residue. Foam-tipped swabs are a necessity as cotton swabs leave behind lint that can potentially cause an overflow in the cuvette.
  • Helps maintain a clean, lint-free cuvette
  • Improves decolorization accuracy
  • Foam tip absorbs residue quickly
  • Quick five-minute procedure
  • 100 swabs per pack