ContactUsLinkComponent
Linearity FLQ TDM for Abbott Systems, AUDIT Microcontrols
Linearity FLQ TDM for Abbott Systems, AUDIT Microcontrols
Catalog # 76485-888
CAS Number:  
undefined
Linearity FLQ TDM for Abbott Systems, AUDIT Microcontrols
Catalog # 76485-888
Supplier Number:  K950M-5
CAS Number:  
Specifications
  • Description:
    Linearity FLQ TDM for Abbott systems
  • Size:
    5 vials
  • Cat. no.:
    76485-888
  • Volume:
    4 ml
Specifications
About this item
The Audit® MicroControls™ Linearity FLQ TDM for Abbott systems is intended to simulate human patient samples for use in determining linearity, calibration verification, and the verification of reportable range for the following analytes: Acetominophen, Amikacin, Carbamezepine, Digoxin, Gentamicin, Phenobarbital, Phenytoin, Salicylate, Theophylline, Tobramycin, Valproic Acid, Vancomycin.
Five distinct levels that demonstrate a linear relationship to each other for their respective analytes, reagents and instruments.