Single-Sample Cryoscope, Model 4250, Advanced Instruments
Product Details & Documents
The single-sample cryoscope, model 4250 is a fast, reliable, and precise system for milk analysis.
- Low maintenance
- On-board printer for better traceability
- Rapid and stable automated calibration
The model 4250 employs the industry preferred freezing point depression (FPD) method for determination of added water to milk. It allows your plant to manage a premium milk supply, ensure quality products and improve your operating efficiency.
Specifications
- Description:Single-sample cryoscope, Model 4250
- Cat. No.:76474-142
- Sample volume:2.0 or 2.5 ml
- Linearity:Less than ±0,5% from straight line (96-wel plate)
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)