Parter Medical Products Petri Dish
76320-910
- Sterility:Sterile
- Description:Petri dish, large
- Size:150×15 mm
- Cat. no.:76320-910
- Packaging:120 per case
The high quality of Parter Medical's Petri dishes is the result of dedication to product excellence, innovative product design and consumer satisfaction.
- Manufactured from pure virgin polystyrene
- Quality design and dependable service