ContactUsLinkComponent
Linearity FD iPTH for Siemens Centaur, AUDIT MircoControls
Linearity FD iPTH for Siemens Centaur, AUDIT MircoControls
Catalog # 76479-168
CAS Number:  
Linearity FD iPTH for Siemens Centaur, AUDIT MircoControls
Catalog # 76479-168
Supplier Number:  K813M-5
CAS Number:  
Specifications
  • Description:
    Linearity FD iPTH for Siemens centaur, 5 vials
  • Size:
    5x 3 ml
  • Cat. no.:
    76479-168
Specifications
About this item
Intended to simulate human patient serum samples for purpose of determining linearity, calibration verification and verification of reportable range for iPTH.
The Linearity FD iPTH for Siemens Centaur is a five level linearity set. Each level contains the following analytes: iPTH. These five levels demonstrate a linear relationship to each other for their respective analytes.