Specifications
- Sample type:StoolWastewaterSoil
- Target molecule:DNA
- Description:FastDNA™ fecal kit
- Size:2 mL tubes
- Storage conditions:15…30° C
- Instrument Compatibility:FastPrep-24™ or FastPrep-96™
- Sample size:Up to 500 mg per preparation
- Cat. no.:75784-608
- Supplier No.:116570200
Specifications
About this item
Designed to quickly and efficiently isolate PCR-ready genomic DNA from fresh or frozen human and animal stool samples.
- Pack size: 50 preparations
- Harndess: Ceramic Spheres, Vickers 800; Silica Spheres, Knoop 515; Glass Bead, Knoop 515
- Specific Gravity: Ceramic Spheres, 4; Silica Spheres, 2.5; Glass Bead, 2.5
Host cells, bacteria, fungi, viruses, protists and other cells present in fecal samples are easily lysed within 40 seconds with any FastPrep® instrument. The FastDNA SPIN kit for Feces contains pre-filled 2 ml Lysing Matrix tubes for the homogenization step as well as all the reagents needed to extract DNA in a purification process eliminating PCR inhibitors.
The Fast DNA® SPIN Kit for Feces quickly and efficiently isolates PCR-ready genomic DNA from fresh or frozen human and animal stool samples.
Kit contents: Lysing matrix E 50×2.0 ml tubes, PLS solution 25 ml, MT buffer 8 ml, sodium phosphate buffer 100 ml, PPS 25 ml, binding matrix 66 ml, wash buffer #1 120 ml, wash buffer #2 12 ml, TES 20 ml, BBS gel loading dye 200 μl, SPIN filter tubes 50 each, catch tubes 50 each.
Reagent: Sodium phosphate buffer - pH range: 7.8 to 8.2
Reagent: MT buffer - pH Range: 5.8 to 6.2
Reagent: PPS protein precipitate solution - pH range: 5.8 to 6.2
Reagent: Binding matrix - pH range: 6.5 to 7.2