Human Recombinant Syndecan-1 (from HEK293), Unconjugated
103889-656
:
- Conjugation:Unconjugated
- Species:Human
- Size:100 µg
- Storage conditions:–20 °C
- Endotoxin content:Less than 1.0 EU per μg by the LAL method.
- Biological activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; 2e5 of anti-Syndecan-1 CAR-293 cells were stained with 100 μL of 0.3 μg/mL of Human Syndecan-1, Fc Tag (Cat. No. SY1-H5253) and negative control protein respectively, washed and then followed by PE anti-human IgG Fc antibody and analyzed with FACS (Routinely tested).
- Gene ID:P18827-1
- Protein synonyms:SDC1,Syndecan-1,CD138,SYND1,SDC
- Protein/peptide name:Syndecan-1
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:50.3 kDa
- Endotoxin level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping temperature:RT
- Cat. no.:103889-656