Order Entry
Canada
ContactUsLinkComponent
Human Recombinant CD27 Ligand (from HEK293), FITC Labeled Protein
Human Recombinant CD27 Ligand (from HEK293), FITC Labeled Protein
Catalog # 103889-634
CAS Number:  
Human Recombinant CD27 Ligand (from HEK293), FITC Labeled Protein
Catalog # 103889-634
Supplier Number:  CDDHF2D4200UG
CAS Number:  

Specifications

  • Species:
    Human
  • Size:
    200 µg
  • Storage conditions:
    –20 °C, aviod light
  • Endotoxin content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4 . Normally trehalose is added as protectant before lyophilization.
  • Gene ID:
    P32970-1
  • Protein synonyms:
    CD70,CD27LG,TNFSF7,TNFSF7G,CD27L
  • Protein/peptide name:
    CD27 Ligand
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular weight:
    49.3 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4 . Normally trehalose is added as protectant before lyophilization.
  • Shipping temperature:
    RT
  • Cat. no.:
    103889-634

Specifications

About this item