Order Entry
Canada
ContactUsLinkComponent
 
Human Recombinant CLEC12A (from HEK293), Precision Avi
Catalog # 103889-626
CAS Number:  
Human Recombinant CLEC12A (from HEK293), Precision Avi
Catalog # 103889-626
Supplier Number:  CLAH82F7200UG
CAS Number:  

Specifications

  • Species:
    Human
  • Size:
    200 µg
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; 2e5 of anti-CLEC12A CAR-293 were stained with 100 μL of 3 μg/mL of Biotinylated Human CLEC12A, Fc,Avitag (Cat. No. CLA-H82F7) and negative control protein respectively, washed and then followed by PE-SA and analyzed with FACS (Routinely tested).
  • Gene ID:
    Q5QGZ9-2;160364;NP_612210.4
  • Protein synonyms:
    CLEC12A,MICL,CLL-1,CLL1,DCAL2,DCAL-2,CD371
  • Protein/peptide name:
    CLEC12A
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    51.9 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping temperature:
    RT
  • Cat. no.:
    103889-626

Specifications

About this item