Human Recombinant M-CSF R (from HEK293), Unconjugated
Catalog # 103889-520
Supplier: ACRO Biosystems
CAS Number:
Specifications
- Conjugation:Unconjugated
- Species:Human
- Size:100 µg
- Storage conditions:–20 °C
- Endotoxin content:Less than 0.1 EU per μg by the LAL method.
- Biological activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Gene ID:P07333-1;1436;NP_005202.2
- Protein synonyms:CSF1R,C-FMS,CD115,CSFR,FIM2,FMS,M-CSFR
- Protein/peptide name:M-CSF R
- Purity:>90% as determined by SDS-PAGE.
- Molecular weight:81.4 kDa
- Endotoxin level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping temperature:RT
- Cat. no.:103889-520