Order Entry
Canada
ContactUsLinkComponent
 
Human Recombinant PLGF (from HEK293), Unconjugated
  103889-466
 :  
Human Recombinant PLGF (from HEK293), Unconjugated
  103889-466
 :  PGFH52H5500UG
 :  

 

  • Conjugation:
    Unconjugated
  • Species:
    Human
  • Size:
    500 µg
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Gene ID:
    P49763-2;5228;NP_001193941.1
  • Protein synonyms:
    PGF,PLGF,PlGF,PGFL,SHGC-10760
  • Protein/peptide name:
    PLGF
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular weight:
    16.6 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping temperature:
    RT
  • Cat. no.:
    103889-466