Order Entry
Canada
ContactUsLinkComponent
Human Recombinant NCAM-1 (from HEK293), Unconjugated
Human Recombinant NCAM-1 (from HEK293), Unconjugated
Catalog # 103889-434
CAS Number:  
Human Recombinant NCAM-1 (from HEK293), Unconjugated
Catalog # 103889-434
Supplier Number:  NC1H5223100UG
CAS Number:  

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    Human
  • Size:
    100 µg
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 500 mM NaCl, pH8.0. Normally trehalose is added as protectant before lyophilization.
  • Gene ID:
    P13591-2;4684;NP_851996.2
  • Protein synonyms:
    CD56,MSK39,NCAM1,N-CAM-1
  • Protein/peptide name:
    NCAM-1
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    79.3 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 500 mM NaCl, pH8.0. Normally trehalose is added as protectant before lyophilization.
  • Shipping temperature:
    RT
  • Cat. no.:
    103889-434

Specifications

About this item