Order Entry
Canada
ContactUsLinkComponent
Human Recombinant SIRP alphaV2 (from HEK293), Precision Avi
Human Recombinant SIRP alphaV2 (from HEK293), Precision Avi
Catalog # 103889-384
CAS Number:  
Human Recombinant SIRP alphaV2 (from HEK293), Precision Avi
Catalog # 103889-384
Supplier Number:  SI2H82W8200UG
CAS Number:  

Specifications

  • Species:
    Human
  • Size:
    200 µg
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 0.1 EU per μg by the LAL method.
  • Biological activity:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Gene ID:
    ATD50864.1
  • Protein synonyms:
    SHPS1,SIRPA,CD172A,BIT,MFR,MYD1,P84,PTPNS1
  • Protein/peptide name:
    SIRP alphaV2
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    40.9 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping temperature:
    RT
  • Cat. no.:
    103889-384

Specifications

About this item