Human Recombinant CEACAM-5 (from HEK293), FITC Labeled Protein
Catalog # 103889-276
Supplier: ACROBIOSYSTEMS INC MS
CAS Number:
Specifications
- Species:Human
- Size:200 µg
- Storage conditions:–20 °C, aviod light
- Endotoxin content:Less than 1.0 EU per μg by the LAL method.
- Biological activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Gene ID:P06731-1
- Protein synonyms:CEACAM-5,CD66e,CEA,Meconium antigen 100
- Protein/peptide name:CEACAM-5
- Purity:>90% as determined by SDS-PAGE.
- Molecular weight:72.2kDa
- Endotoxin level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping temperature:RT
- Cat. no.:103889-276