Human Recombinant ENPP3 (from HEK293), Unconjugated
Catalog # 103889-188
Supplier: ACROBIOSYSTEMS INC MS
CAS Number:
Specifications
- Conjugation:Unconjugated
- Species:Human
- Size:100 µg
- Storage conditions:–20 °C
- Endotoxin content:Less than 1.0 EU per μg by the LAL method.
- Biological activity:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH8.0. Normally trehalose is added as protectant before lyophilization.
- Gene ID:O14638-1;5169;NP_005012.2
- Protein synonyms:ENPP3,CD203c,NPP3,E-NPP3,PD-Ibeta,NPPase,PDNP3
- Protein/peptide name:ENPP3
- Purity:>90% as determined by SDS-PAGE.
- Molecular weight:96.9 kDa
- Endotoxin level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH8.0. Normally trehalose is added as protectant before lyophilization.
- Shipping temperature:RT
- Cat. no.:103889-188